Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-Tyrosine Antibody (P-TYR-01), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB500335FR
Description
Phospho-Tyrosine Monoclonal specifically detects Phospho-Tyrosine in Non-species specific samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
Phospho-Tyrosine | |
Monoclonal | |
DyLight 680 | |
2 amino 3(4 hydroxyphenyl) propanoic acid, 4 hydroxyphenylalanine, phosphotyrosine, pTyrosine, Tyrosine | |
Phosphotyrosine conjugated to BSA. | |
0.1 mL | |
Primary | |
All species | |
IgG1 |
Western Blot, Immunocytochemistry, Immunofluorescence | |
P-TYR-01 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
Mouse | |
Affinity Purified | |
RUO | |
The antibody P-Tyr-01 detects tyrosine phosphorylation in activated cells. | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction