Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-Tyrosine Antibody (PY20), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP233166MFV500
Description
Phospho-Tyrosine Monoclonal antibody specifically detects Phospho-Tyrosine in Non-species specific samples. It is validated for Flow CytometrySpecifications
Phospho-Tyrosine | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Store at 4C in the dark. | |
IgG2b |
Flow Cytometry | |
PY20 | |
Flow Cytometry | |
2 amino 3(4 hydroxyphenyl) propanoic acid, 4 hydroxyphenylalanine, phosphotyrosine, pTyrosine, pY20, Tyrosine | |
Phosphotyrosine conjugated to KLH | |
0.1 mL | |
Neuroscience, Phospho Specific, Signal Transduction, Tyrosine Kinases | |
Non-species specific | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction