Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-Tyrosine Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB1676100
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1676100 100 μg
MAB1676SP 25 μg
MAB1676500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB1676100 Supplier R&D Systems Supplier No. MAB1676100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Phospho-Tyrosine Monoclonal specifically detects Phospho-Tyrosine in Multi-Species samples. It is validated for Western Blot, Simple Western, Immunoprecipitation, Immunocytochemistry.

Specifications

Antigen Phospho-Tyrosine
Applications Western Blot, Immunoprecipitation, Immunocytochemistry
Classification Monoclonal
Clone 179003
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Simple Western 50 ug/mL, Immunoprecipitation 5 ug/500 ug cell lysate, Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Alias 2 amino 3(4 hydroxyphenyl) propanoic acid, 4 hydroxyphenylalanine, PhosphoTyrosine, pTyrosine, Tyrosine
Host Species Mouse
Immunogen KLH-coupled phospho-tyrosine synthetic peptide
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Detects proteins containing phosphorylated tyrosine residues. ELISA and Western blot analyses using pervanadate-treated cell lysates indicate that clone 179003 binds phospho-tyrosine in a broad manner largely independent of the surrounding amino acid sequence. No cross-reactivity with proteins or peptides containing phosphorylated serine or threonine residues is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Multi-species
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.