Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-Tyrosine Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB16761
This item is not returnable.
View return policy
Description
Phospho-Tyrosine Monoclonal specifically detects Phospho-Tyrosine in Multi-Species samples. It is validated for Western Blot, Immunocytochemistry.Specifications
Phospho-Tyrosine | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL | |
2 amino 3(4 hydroxyphenyl) propanoic acid, 4 hydroxyphenylalanine, PhosphoTyrosine, pTyrosine, Tyrosine | |
KLH-coupled Phospho-Tyrosine synthetic peptide | |
500 μg | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Western Blot, Immunocytochemistry | |
216954 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Detects proteins containing phosphorylated tyrosine residues in Western blots. In Western blots of pervanadate-treated cell lysates, clone 216954 binds Phospho-Tyrosine in a broad manner largely independent of the surrounding amino acid sequence. No cross-reactivity with proteins or peptides containing phosphorylated serine or threonine residues is observed. | |
Multi-species | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction