Learn More
Invitrogen™ Phospho-Tyrosine Hydroxylase (Ser62) Polyclonal Antibody

Description
This antibody is predicted to react with mouse, rat, and monkey based on sequence homology.
Specifications
Specifications
| Antigen | Phospho-Tyrosine Hydroxylase (Ser62) |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | TH |
| Gene Accession No. | P04177, P07101, P24529 |
| Gene Alias | dystonia 14; DYT14; DYT5b; EC 1.14.16.2; HGNC:11782; Th; TH isoform 3; TH isoform a; th1; TH-4; The; TY3H; TYH; TYH antibody; Tyrosine 3-hydroxylase; tyrosine 3-monooxygenase; tyrosine hydroxylase |
| Gene Symbols | TH |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.