Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-UBF-1 (Thr117) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5105512
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5105512 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5105512 Supplier Invitrogen™ Supplier No. PA5105512
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of UBF1 only when phosphorylated at Thr117.

UBF-1 encodes a member of the HMG-box DNA-binding protein family. The encoded protein plays a critical role in ribosomal RNA transcription as a key component of the pre-initiation complex, mediating the recruitment of RNA polymerase I to rDNA promoter regions. The encoded protein may also play important roles in chromatin remodeling and pre-rRNA processing, and its activity is regulated by both phosphorylation and acetylation. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene. Pseudogenes of this gene are located on the short arm of chromosomes 3, 11 and X and the long arm of chromosome 11.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-UBF-1 (Thr117)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene UBTF
Gene Accession No. P17480, P25976, P25977
Gene Alias 90-kDa nucleolus organizer region autoantigen; A930005G04Rik; Autoantigen NOR-90; NOR90; NOR-90; Nucleolar transcription factor 1; Tcfubf; transcription factor UBF; UBF; UBF1; UBF-1; UBF2; Ubtf; upstream binding transcription factor, RNA polymerase I; upstream-binding factor 1
Gene Symbols UBTF
Host Species Rabbit
Immunogen A synthesized peptide derived from human UBTF(Accession P17480), corresponding to amino acid residues around phosphorylated Thr117.
Purification Method Sequential chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21429, 25574, 7343
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.