Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Phospho-ULK1 (Ser555) Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA578249
Description
Positive Control: MCF-7, MCF-7 (100 μM CCCP treatment for 2 hr) Predicted Reactivity: Mouse (100%), Rat (100%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
ULK1 is a serine/threonine protein kinase that plays critical role during initial stages of autophagy which is a vital response to nutrient starvation. The conserved C-terminal domain (CTD) of ULK1 controls the regulatory function and localization of the protein. Knockdown of ULK1 inhibits the autophagic response as well as inhibiting rapamycin-induced autophagy consistent with a role downstream of mTOR. ULK1 forms a complex with FIP200 and ATG13 and this complex is essential for starvation-induced autophagy. Both FIP200 and ATG13 are critical for correct localization of ULK1 to the pre-autophagosome and stability of ULK1 protein. ULK1 is phosphorylated by the mTOR pathway in a nutrient starvation-regulated manner.
Specifications
Phospho-ULK1 (Ser555) | |
Polyclonal | |
Unconjugated | |
ULK1 | |
APG1; ATG 1; ATG1; ATG1 autophagy related 1 homolog; ATG1A; AU041434; Autophagy-related protein 1 homolog; C. elegans; EC:2.7.11.1; FLJ38455; hATG1; KIAA0722; mKIAA0722; Serine/threonine-protein kinase ULK1; serine/threonine-protein kinase Unc51.1; ULK 1; Ulk1; UNC 51; UNC51; unc-51 like autophagy activating kinase 1; unc-51 like kinase 1; Unc51.1; unc-51-like kinase 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
8408 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 | |
O75385 | |
ULK1 | |
Recombinant protein encompassing a sequence within the center region of human ULK1 (phospho Ser555). The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction