Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-ULK1 (Ser758) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5105130
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5105130 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5105130 Supplier Invitrogen™ Supplier No. PA5105130
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of ULK1 only when phosphorylated at Ser757 of mouse ULK1 or Ser758 of human ULK1.

ULK1 is a serine/threonine protein kinase that plays critical role during initial stages of autophagy which is a vital response to nutrient starvation. The conserved C-terminal domain (CTD) of ULK1 controls the regulatory function and localization of the protein. Knockdown of ULK1 inhibits the autophagic response as well as inhibiting rapamycin-induced autophagy consistent with a role downstream of mTOR. ULK1 forms a complex with FIP200 and ATG13 and this complex is essential for starvation-induced autophagy. Both FIP200 and ATG13 are critical for correct localization of ULK1 to the pre-autophagosome and stability of ULK1 protein. ULK1 is phosphorylated by the mTOR pathway in a nutrient starvation-regulated manner.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-ULK1 (Ser758)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ULK1
Gene Accession No. O70405, O75385
Gene Alias APG1; ATG 1; ATG1; ATG1 autophagy related 1 homolog; ATG1A; AU041434; Autophagy-related protein 1 homolog; C. elegans; EC:2.7.11.1; FLJ38455; hATG1; KIAA0722; mKIAA0722; Serine/threonine-protein kinase ULK1; serine/threonine-protein kinase Unc51.1; ULK 1; Ulk1; UNC 51; UNC51; unc-51 like autophagy activating kinase 1; unc-51 like kinase 1; Unc51.1; unc-51-like kinase 1
Gene Symbols ULK1
Host Species Rabbit
Immunogen A synthesized peptide derived from mouse ULK1(Accession O75385), corresponding to amino acid residues around phosphorylated Ser758.
Purification Method Sequential chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22241, 360827, 8408
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.