Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-VANGL2 (Thr78, Ser79) Recombinant Rabbit Monoclonal Antibody (5K4D6)
GREENER_CHOICE

Catalog No. PIMA542958
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542958 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542958 Supplier Invitrogen™ Supplier No. MA542958
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

The target is usually found in the following locations: Cell membrane, Multi-pass membrane protein.

Van Gogh is an alternate name for the Drosophila strabismus protein. Van Gogh like-2 (VANGL2) is a 521 amino acidprotein in vertebrates similar to strabismus 1 of Drosophila and a component of the frizzled-disheveled tissue polarity pathway. It interacts through it's C-terminal region with the N-terminal half of DVL1, DVL2 and DVL3 and is a potent tumor suppressor. It ispredominantly involved in the control of early morphogenesis and patterning of both axial midline structures and the development of neural plate. It plays a crucial role in the regulation of planar cell polarity, particularly in the orientation of stereociliary bundles in the cochlea. VANGL2 is required for polarization and movement of myocardializing cells and seems to act via RHOA signaling to regulate this process. It is expressed in a wide range of tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-VANGL2 (Thr78, Ser79)
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 5K4D6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Vangl2
Gene Accession No. Q9ULK5
Gene Alias C530001F03Rik; KIAA1215; loop tail associated protein; loop-tail; Loop-tail protein 1; loop-tail protein 1 homolog; Loop-tail-associated protein; Lp; LPP1; Ltap; MGC119403; MGC119404; RGD:1309442}; ska>m17Jus>; ska17; STB1; stbm; STBM1; strabismus; Strabismus 1; van Gogh-like protein 2; vang; vang (van gogh)-like 2; Vang1l2; VANGL planar cell polarity protein 2; Vangl2; vangl2 {ECO:0000312; vang-like 2 (van gogh, Drosophila); vang-like protein 2
Gene Symbols Vangl2
Host Species Rabbit
Immunogen A phospho specific peptide corresponding to residues surrounding T78/S79 of human VANGL2.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57216
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.