Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-VAV1 (Tyr174) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA536699
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA536699 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA536699 Supplier Invitrogen™ Supplier No. PA536699
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 100 kDa. Purity is >95% by SDS-PAGE.

The VAV1 gene, located on chromosome 19p13.2, encodes for a protein that functions as a guanine nucleotide exchange factor (GEF). Structurally, the VAV1 protein contains several functional domains, including Dbl homology (DH) and pleckstrin homology (PH) domains, which facilitate its role in exchange activity, and SH3 and SH2 domains, critical for signal transduction. VAV1 is primarily expressed in hematopoietic cells and plays a crucial role in the regulation of immune responses, particularly in T-cell receptor signaling. It influences actin cytoskeleton reorganization, cell proliferation, and differentiation, making it vital for proper immune system function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-VAV1 (Tyr174)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene VAV1
Gene Accession No. P15498, P54100
Gene Alias oncogene vav; p95; p95vav; Proto-oncogene vav; VAV; vav 1 guanine nucleotide exchange factor; vav 1 oncogene; vav guanine nucleotide exchange factor 1; vav oncogene; vav proto-oncogene; VAV1; vav-T
Gene Symbols VAV1
Host Species Rabbit
Immunogen Synthetic phosphopeptide derived from human VaV1 around the phosphorylation site of Tyrosine 174.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25156, 7409
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.