Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-VAV3 (Tyr141) Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501980784
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-0784 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-0784 Supplier Bioss Supplier No. BS10271R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene is a member of the VAV gene family. The VAV proteins are guanine nucleotide exchange factors (GEFs) for Rho family GTPases that activate pathways leading to actin cytoskeletal rearrangements and transcriptional alterations. This gene product acts as a GEF preferentially for RhoG, RhoA, and to a lesser extent, RAC1, and it associates maximally with the nucleotide-free states of these GTPases. Alternatively spliced transcript variants encoding different isoforms have been described for this gene.

Specifications

Antigen Phospho-VAV3 (Tyr141)
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene VAV3
Gene Accession No. Q9UKW4
Gene Alias A530094I06Rik; AA986410; FLJ40431; Guanine nucleotide exchange factor VAV3; Idd18.1; vav 3 guanine nucleotide exchange factor; vav 3 oncogene; vav guanine nucleotide exchange factor 3; VAV3; VAV-3
Gene Symbols VAV3
Host Species Rabbit
Immunogen KLH conjugated synthetic phosphopeptide derived from human VAV3 around the phosphorylation site of Tyr141.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10451
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.