Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ phospho-VE-Cadherin (Tyr685), Rabbit, Alexa Fluor 555, Polyclonal,

Catalog No. ABT1760ALMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Alexa Fluor 555
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ABT1760ALMI 100 μL Alexa Fluor 555
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABT1760ALMI Supplier MilliporeSigma™ Supplier No. ABT1760AF555
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Cadherin-5 (UniProt P33151; also known as 7B4 antigen, CD144, Vascular endothelial cadherin, VE-Cad, VE-cadherin) is encoded by the CDH5 gene (Gene ID 1003) in human. Cadherins (calcium-dependent adhesion) are type-1 transmembrane proteins that form adherens junctions in tissues and play important roles in mediating cell adhesion. Cadherins are designated with a prefix that specifies their tissue association. Vascular endothelial-cadherin (VE-cadherin) is the transmembrane component of the endothelial adherens junction between vascular endothelial cells (ECs) and plays a pivotal role in endothelium integrity and in the control of vascular permeability. One characteristic of VEGF-induced vascular permeability is the phosphorylation of VE-cadherin by Src family kinases (SFK), which leads to VE-cadherin internalization and the destabilization of adherens junctions. Likewise, VE-cadherin overexpression is shown to decrease the permeability of endothelial monolayers in vitro. SFK activation by dominant negative Csk overexpression is reported to result in VE-cadherin phosphorylation at tyrosines 658, 685, and 731 in human dermal microvascular endothelial cells (HDMECs) without affecting HDMEC monolayer permeability. On the other hand, expression of constitutively active Src promoted VE-cadherin phosphorylation on tyrosines 658 and 731 and decreased barrier function, suggesting that concurrent signaling events in addition to VE-cadherin tyrosine phosphorylation are needed to promote barrier permeability in response to inflammatory mediators or growth factors. VE-cadherin is initially produced with a signal peptide (a.a. 1-24) and a propeptide (a.a. 25-45) sequence, the removal of which yields tthe mature protein containing a large extracellular region (a.a. 46-599) with five cadherine repeats (a.a. 46-149, 150-256, 257-371, 372-476, 477-593), followed by a transmembrane segment (a.a. 600-620) and a cytoplasmic domain (a.a. 621-784).
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-VE-Cadherin (Tyr685)
Applications Immunocytochemistry
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Alexa Fluor 555
Formulation Purified rabbit polyclonal antibody Alexa Fluor™ 555 conjugate in PBS with 1.5% BSA and 0.05% Sodium Azide.
Gene CDH5
Gene Accession No. P33151
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to a sequence surrounding phosphorylated Tyr685 of human VE-cadherin isoform 1 precursor sequence.
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cell Structure
Primary or Secondary Primary
Gene ID (Entrez) NP_001786
Test Specificity Target specificity of the unconjugated antibody (Mfr. No. ABT1760) has been verified by immunogen peptide blocking in Western blotting application. Numbering of the target VE-cadherin phosphotyrosine residue varies depending on whether the N-terminal signal peptide sequence and propeptide sequence are included, as well as the species and spliced isoform a particular numbering is based on. Target pTyr this rabbit polyclonal detects corresponds to pTyr685/pTyr638 of human spliced isoform 1 precursor/mature sequence (UniProt P33151-1), pTyr570/pTyr523 of human spliced isoform 2 precursor/mature sequence (UniProt P33151-2), pTyr685/pTyr640 of bovine precursor/mature sequence (UniProt Q6URK6), and pTyr684/pTyr40 of porcine precursor/mature sequence (UniProt O02840). Equivalent target site sequence is not present in mouse, rat, or chicken VE-cadherin sequence.
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.