Learn More
phospho-VE-cadherin (Tyr731) Rabbit anti-Human, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | phospho-VE-cadherin (Tyr731) |
| Applications | Western Blot, Inhibition Assays |
| Classification | Polyclonal |
| Concentration | Please refer to lot specific datasheet. |
| Conjugate | Unconjugated |
| Gene Symbols | CDH5 |
| Host Species | Rabbit |
| Immunogen | KLH-conjugated linear peptide corresponding to 11 amino acids surrounding phosphorylated tyrosine 731 from human Cadherin-5. |
| Purification Method | Affinity Purified |
| Quantity | 200 μL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.