Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Phospho-VEGFR1/Flt-1 (Y1213) Antibody, R&D Systems™

Rabbit Polyclonal Antibody has been used in 11 publications
Supplier: R&D Systems AF4170SP
This item is not returnable.
View return policy
Description
VEGFR1/Flt-1 Polyclonal specifically detects VEGFR1/Flt-1 in Human samples. It is validated for Western Blot.Specifications
VEGFR1/Flt-1 (p tyr1213) | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
FLT1 | |
Phosphopeptide containing human VEGFR1/Flt-1 Y1213 site | |
25 μg | |
Primary | |
Detects human VEGFR1/Flt-1 when phosphorylated at Y1213. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
EC 2.7.10, EC 2.7.10.1, FLT, FLT1, FLT-1, Fms-like tyrosine kinase 1, fms-related tyrosine kinase 1 (vascular endothelial growth factor/vascularpermeability factor receptor), FRT, Tyrosine-protein kinase FRT, Tyrosine-protein kinase receptor FLT, vascular endothelial growth factor receptor 1, Vascular permeability factor receptor, VEGF R1, VEGFR1, VEGFR-1 | |
Rabbit | |
Affinity Purified | |
RUO | |
2321 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction