Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-XIRP1 (Ser295) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114674
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114674 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114674 Supplier Thermo Scientific Supplier No. PA5114674
Add to Cart
Add to Cart

Phospho-XIRP1 (Ser295) Polyclonal Antibody for Western Blot, IHC (PFA)

TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-XIRP1 (Ser295)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene XIRP1
Gene Accession No. O70373, Q702N8
Gene Alias AI415219; cardiomyopathy associated 1; cardiomyopathy-associated protein 1; CMYA1; LOC100910104; mXin alpha; Xin; xin actin binding repeat containing 1; xin actin-binding repeat containing 1; xin actin-binding repeat-containing protein 1; xin actin-binding repeat-containing protein 1-like; Xin alpha; XIRP1
Gene Symbols XIRP1
Host Species Rabbit
Immunogen A synthesized peptide derived from mouse XIRP1 around the phosphorylation site of Ser295.
Purification Method Sequential Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100910104, 165904, 22437
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.