Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-YAP1 (Ser127) Recombinant Rabbit Monoclonal Antibody (SN0718)
GREENER_CHOICE

Catalog No. PIMA541140
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA541140 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA541140 Supplier Invitrogen™ Supplier No. MA541140
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Phospho-YAP1 (Ser127) Recombinant Monoclonal Antibody for IHC (P), Western Blot

YAP (Yes-Associated Protein) is a founding member of the WW domain family of proteins, transcriptional co-activator and oncogene. Its gene is transcriptionally activated in squamous cell carcinoma cells. YAP can act both as a coactivator and corepressor and is the critical downstream regulatory target in the Hippo signaling pathway. The Hippo signaling pathway plays a pivotal role in organ size control and tumour suppression by restricting proliferation and promoting apoptosis. Also plays key roles in tissue tension, 3D tissue shape, cell proliferation, cell death, and cell migration. Mutations in the IRF5 gene results in coloboma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-YAP1 (Ser127)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone SN0718
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene YAP1
Gene Accession No. P46937, P46938, Q2EJA0
Gene Alias 65 kDa Yes-associated protein; AI325207; cb194; CH211-181P1.5-001; COB1; Protein yorkie homolog; sb:cb194; si:ch211-181p1.5; si:dkey-3b8.3; transcriptional coactivator YAP1; wu:fc18c04; yap; Yap1; YAP2; Yap65; Yes associated protein 1; Yes1 associated transcriptional regulator; yes-associated protein 1; yes-associated protein 2; yes-associated protein YAP65 homolog; yes-associated protein, 65 kDa; Yki; Yorkie; yorkie homolog; zgc:158380; zYAP
Gene Symbols YAP1
Host Species Rabbit
Immunogen Synthetic phospho-peptide corresponding to residues surrounding Ser127 of human YAP1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10413, 22601, 363014
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.