Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Phospho-YBX1 (Ser102) Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA537669
Description
A suggested positive control for Western blot is HepG2 cells.
YBX1 binds to splice sites in pre-mRNA and regulates splice site selection. This protein binds and stabilizes cytoplasmic mRNA and contributes to the regulation of translation by modulating the interaction between the mRNA and eukaryotic initiation factors. It binds to promoters that contain a Y-box (5'-CTGATTGGCCAA-3'), such as HLA class II genes. It regulates the transcription of numerous genes and promotes separation of DNA strands that contain mismatches or are modified by cisplatin. It has endonucleolytic activity and can introduce nicks or breaks into double-stranded DNA (in vitro), and it may play a role in DNA repair.
Specifications
| Phospho-YBX1 (Ser102) | |
| Polyclonal | |
| Unconjugated | |
| YBX1 | |
| 1700102N10Rik; BP8; BP-8; Byb1; C79409; Cbfa; CBF-A; CCAAT-binding; CCAAT-binding transcription factor 1 subunit A; CCAAT-binding transcription factor I subunit A; CSDA2; CSDB; dbpB; DNA binding protein B; DNA-binding protein B; EF1A; EFI-A; enhancer factor 1 alpha; enhancer factor 1 subunit A; enhancer factor I subunit A; major histocompatibility complex, class II, Y box-binding protein I; MDR NF1; MDR-NF1; MGC104858; MGC110976; MGC117250; Msy1; Msy-1; mYB-1a; Nsep1; NSEP-1; nuclease sensitive element binding protein 1; nuclease-sensitive element-binding protein 1; Y box binding protein 1; Y box protein 1; Y box transcription factor; YB1; YB-1; Y-box binding protein 1; Y-box transcription factor; Y-box-binding protein 1; YBX1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 22608, 4904, 500538 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P62960, P62961, P67809 | |
| YBX1 | |
| Peptide sequence around phosphorylation site of Serine 102(L-R-S(p)-V-G) derived from Human YB1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction