Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-ZAK (Ser165) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564615
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564615 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564615 Supplier Invitrogen™ Supplier No. PA564615
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Phospho-ZAK (Ser165) Polyclonal Antibody detects endogenous levels of ZAK only when phosphorylated at Ser165.

ZAK is a leucine zipper-containing mitogen-activated protein related to the MLKs in both sequence and function. Like the MLKs and other MAP3Ks, ZAK activates MAP2Ks. ZAK is a member of the MAPKKK family of signal transduction molecules and encodes a protein with an N-terminal kinase catalytic domain, followed by a leucine zipper motif and a sterile-alpha motif. This magnesium-binding protein forms homodimers and is located in the cytoplasm. The protein mediates gamma radiation signaling leading to cell cycle arrest and activity of this protein plays a role in cell cycle checkpoint regulation in cells. The protein also has pro-apoptotic activity. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-ZAK (Ser165)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
Gene MAP3K20
Gene Accession No. Q9ESL4, Q9NYL2
Gene Alias AV006891; AZK; B230120H23Rik; HCCS4; HCCS-4; Human cervical cancer suppressor gene 4 protein; leucine zipper- and sterile alpha motif kinase ZAK; Leucine zipper- and sterile alpha motif-containing kinase; Map3k20; Mitogen-activated protein kinase kinase kinase 20; Mitogen-activated protein kinase kinase kinase MLT; mixed lineage kinase 7; mixed lineage kinase with a leucine zipper and a sterile alpha motif; mixed lineage kinase-related kinase; mixed lineage kinase-related kinase MRK-beta; MLK7; mlklak; MLK-like mitogen-activated protein triple kinase; MLK-related kinase; MLT; MLTK; MLTKalpha; MLTKbeta; MRK; pk; RGD1561394; SFMMP; sterile alpha motif and leucine zipper containing kinase AZK; sterile alpha motif- and leucine zipper-containing kinase AZK; sterile-alpha motif and leucine zipper containing kinase AZK; ZAK
Gene Symbols MAP3K20
Host Species Rabbit
Immunogen A synthesized peptide derived from human ZAK (Phospho-Ser165).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51776, 65964
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.