Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Phospho-ZCWCC1 (Ser615) Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5114606
Description
Antibody detects endogenous levels of MORC2 only when phosphorylated at Ser615.
Exhibits a cytosolic function in lipogenesis, adipogenic differentiation, and lipid homeostasis by increasing the activity of ACLY, possibly preventing its dephosphorylation. May act as a transcriptional repressor. Down-regulates CA9 expression.
Specifications
| Phospho-ZCWCC1 (Ser615) | |
| Polyclonal | |
| Unconjugated | |
| MORC2 | |
| 8430403M08Rik; ATPase MORC2; ATPase MORC2A; CMT2Z; Kiaa0852; microrchidia 2A; MORC family CW-type zinc finger 2; MORC family CW-type zinc finger protein 2; MORC family CW-type zinc finger protein 2A; MORC2; Morc2a; ZCW3; ZCWCC1; zinc finger CW-type coiled-coil domain protein 1; zinc finger, CW-type with coiled-coil domain 1 | |
| Rabbit | |
| Sequential Chromatography | |
| RUO | |
| 22880 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| Q9Y6X9 | |
| MORC2 | |
| A synthesized peptide derived from human MORC2 around the phosphorylation site of Ser615. | |
| 100 μL | |
| Primary | |
| Human, Monkey | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction