Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-ZEB2 (Ser784) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114677
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114677 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114677 Supplier Invitrogen™ Supplier No. PA5114677
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of ZEB2 only when phosphorylated at Ser784.

ZEB2, initially identified as Smad interacting-protein 1, is normally located in the nucleus and functions as a DNA-binding transcriptional repressor that interacts with activated SMADs. Like the homologous ZEB1, ZEB2 inhibits the transcription of the E-cadherin gene and induces epithelial-mesenchymal transition, a genetic program controlling cell migration during embryonic development and wound healing, in vitro. ZEB2 can also protect cells from DNA damage-induced apoptosis, suggesting that its expression may contribute to tumor progression. Recent evidence has shown that ZEB2 is often observed in the cytoplasm in numerous cancer tissues, indicating that its localization may be regulated in normal and tumor tissues. Mutations in this gene are also associated with Hirschsprung disease/Mowat-Wilson syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-ZEB2 (Ser784)
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene Zeb2
Gene Accession No. O60315, Q9R0G7
Gene Alias 9130203F04Rik; D130016B08Rik; HRIHFB2411; HSPC082; KIAA0569; SIP1; SIP-1; SMAD interacting protein 1; smad-interacting protein 1; SMADIP1; Zeb2; ZFHX1B; Zfx1b; Zfxh1b; zinc finger E-box binding homeobox 2; zinc finger E-box-binding homeobox 2; zinc finger homeobox 1b; zinc finger homeobox protein 1b
Gene Symbols Zeb2
Host Species Rabbit
Immunogen A synthesized peptide derived from human ZEB2 around the phosphorylation site of Ser784.
Purification Method Sequential Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24136, 311071, 9839
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.