Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-ZFP36L1 (Ser334) Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982079
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2079 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2079 Supplier Bioss Supplier No. BS12255R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RNA polymerase (pol) III synthesizes tRNA, 5s rRNA, 7SL RNA and U6 snRNA and is overexpressed in many transformed cell lines and tumors in vivo, since cells must duplicate its protein components before division. Therefore, in order to maintain rapid growth, cells must produce a high level of Pol III transcribed RNA, which requires the presence of the TFIIIB and TFIIIC2 transcription factor complexes. The TFIIIC2 complex is composed of five subunits, TFIIIC220, TFIIIC110, TFIIIC102, TFIIIC90 and TFIIIC63, that are overexpressed in adenovirus transformed cells as well as in malignant cells in vivo, such as ovarian carcinomas. TFIIIC2 recruits RNA pol III and TFIIIB to promoter elements and may be a key component in the deregulation of malignant cells. The TFIIIB complex includes the TATA-binding protein (TBP), TFIIB-related factor 1 (TFIIIB90, BRF1) and TFIIIB, the expression of which are also upregulated in transformed cells. In many carcinomas, the tumor suppressors retinoblastoma (RB) and p53 are inactivated, which affects their ability to bind and inactivate the function of TFIIIB.

Specifications

Antigen Phospho-ZFP36L1 (Ser334)
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene BRF1
Gene Accession No. Q92994
Gene Alias 2510002F24Rik; B - related factor 1; B-related factor 1; BRF; BRF1; BRF-1; BRF1 homolog, subunit of RNA polymerase III transcription initiation factor IIIB; BRF1 homolog, subunit of RNA polymerase III transcription initiation factor IIIB (S. cerevisiae); BRF1, RNA polymerase III transcription initiation factor 90 kDa subunit; BRF1, RNA polymerase III transcription initiation factor 90 subunit; CFDS; epididymis secretory sperm binding protein Li 76p; general transcription factor IIIB, 90kD subunit; GTF3B; hBRF; HEL-S-76p; hTFIIIB90; mTFIIIB90; PCF4; TAF3B2; TAF3C; TAFIII90; TATA box binding protein (TBP)-associated factor 3C; TATA box binding protein (TBP)-associated factor, RNA polymerase III, GTF3B subunit 2; TATA box-binding protein-associated factor, RNA polymerase III, subunit 2; TBP - associated factor, RNA polymerase III, 90kD; TDS4; TF3B90; TFIIIB; TFIIIB90; Transcription factor IIIB 70 kDa subunit; Transcription factor IIIB 90 kDa subunit; transcription factor TFIIIB subunit BRF1; YGR246C; ZFP36L1; zinc finger protein 36, C3H type-like 1
Gene Symbols BRF1
Host Species Rabbit
Immunogen KLH conjugated synthetic phosphopeptide derived from human ZFP36L1 around the phosphorylation site of Ser334.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2972
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.