Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PHOSPHO1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584679
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584679 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584679 Supplier Invitrogen™ Supplier No. PA584679
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RDLSAIYEAI PLSPGMSDLL QFVAKQGACF EVILISDANT FGVESSLRAA GHHSLFRRIL SNPSGPDARG LLALRPF Highest antigen sequence indentity to the following orthologs: Mouse - 90%, Rat - 90%.

Phosphatase that has a high activity toward phosphoethanolamine (PEA) and phosphocholine (PCho). Involved in the generation of inorganic phosphate for bone mineralization. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen PHOSPHO1
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PHOSPHO1
Gene Accession No. Q8TCT1
Gene Alias D11Moh36; phosphatase, orphan 1; PHOSPHO1; Phosphoethanolamine/phosphocholine phosphatase; Phospo1
Gene Symbols PHOSPHO1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PHOSPHO1. Recombinant protein control fragment (Product #RP-108505).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 162466
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.