Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PHOSPHO1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5116152
Description
Antibody detects endogenous levels of total Phospho1.
Phosphatase that has a high activity toward phosphoethanolamine (PEA) and phosphocholine (PCho). Involved in the generation of inorganic phosphate for bone mineralization. [UniProt].
Specifications
| PHOSPHO1 | |
| Polyclonal | |
| Unconjugated | |
| PHOSPHO1 | |
| D11Moh36; phosphatase, orphan 1; PHOSPHO1; Phosphoethanolamine/phosphocholine phosphatase; Phospo1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 162466, 237928, 287644 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| Q8R2H9, Q8TCT1 | |
| PHOSPHO1 | |
| A synthesized peptide derived from human PHOSPHO1(Accession Q8TCT1), corresponding to amino acid residues F195-A245. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction