Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phosphodiesterase 4A/PDE4A Antibody (OTI1C8), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273376MFV500
Description
Phosphodiesterase 4A/PDE4A Monoclonal antibody specifically detects Phosphodiesterase 4A/PDE4A in Human, Rat samples. It is validated for Flow CytometrySpecifications
Phosphodiesterase 4A/PDE4A | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Rat | |
Purified |
Flow Cytometry | |
OTI1C8 | |
Flow Cytometry | |
cAMP-specific 3'-5'-cyclic phosphodiesterase 4A, DPDE2cAMP-specific phosphodiesterase, EC 3.1.4, EC 3.1.4.17, PDE4, PDE46, phosphodiesterase 4A, cAMP-specific, phosphodiesterase 4A, cAMP-specific (dunce, phosphodiesterase 4A, cAMP-specific (dunce (Drosophila)-homologphosphodiesterase E2), phosphodiesterase E2 dunce homolog, Drosophila, phosphodiesterase isozyme 4 | |
Full length human recombinant protein of human PDE4A (NP_006193) produced in HEK293T cell. | |
0.1 mL | |
Apoptosis, Phospho Specific, Signal Transduction, Stem Cell Markers | |
5141 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction