Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phosphodiesterase 4D Antibody (SR2287), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | Phosphodiesterase 4D |
---|---|
Clone | SR2287 |
Dilution | Western Blot 1:500-1:2000 |
Applications | Western Blot |
Classification | Monoclonal |
Description
Phosphodiesterase 4D Monoclonal antibody specifically detects Phosphodiesterase 4D in Human samples. It is validated for Western BlotSpecifications
Phosphodiesterase 4D | |
Western Blot 1:500-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
cAMP-specific (dunce (Drosophila)-homologphosphodiesterase E3), cAMP-specific 3'-5'-cyclic phosphodiesterase 4D, cAMP-specific phosphodiesterase PDE4D6, DKFZp686M11213, Drosophila), dunce-like phosphodiesterase E3, EC 3.1.4, FLJ97311, HSPDE4D, phosphodiesterase 4D, cAMP-specific, phosphodiesterase 4D, cAMP-specific (phosphodiesterase E3 dunce homolog | |
A synthesized peptide derived from human phosphodiesterase 4D (Uniprot #: Q08499) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR2287 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Lipid and Metabolism, Signal Transduction | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
5144 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title