Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phosphoglucomutase 5 Antibody (PGM5/3552) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314132
This item is not returnable.
View return policy
Description
Phosphoglucomutase 5 Monoclonal antibody specifically detects Phosphoglucomutase 5 in Human, Mouse, Rat, Bovine, Rabbit samples. It is validated for Western Blot, ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Protein ArraySpecifications
| Phosphoglucomutase 5 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1 to 2 μg/mL, ELISA, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Immunohistochemistry-Paraffin 1 to 2 μg/mL, Protein Array | |
| aciculin, EC 5.4.2.2, PGM-RP, PGMRPphosphoglucomutase-like protein 5, phosphoglucomutase 5, Phosphoglucomutase-related protein | |
| Recombinant full-length human Phosphoglucomutase 5 protein (Uniprot: Q15124) | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat, Bovine, Rabbit | |
| Purified |
| Western Blot, ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Peptide Array | |
| PGM5/3552 | |
| Unconjugated | |
| 10mM PBS without Preservative | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 5239 | |
| Store at -20 to -80°C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction