Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phosphoglucomutase 5 Mouse anti-Human, Clone: rPGM5/7164, Novus Biologicals™

Mouse Monoclonal Antibody
$240.00 - $492.50
Specifications
Antigen | Phosphoglucomutase 5 |
---|---|
Clone | rPGM5/7164 |
Concentration | 0.2 mg/ml |
Dilution | Western Blot 1-2 ug/ml, ELISA, Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin 2-4 ug/ml |
Applications | Western Blot, ELISA, Immunofluorescence, Immunohistochemistry (Paraffin) |
Description
Phosphoglucomutase 5 Monoclonal antibody specifically detects Phosphoglucomutase 5 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
Phosphoglucomutase 5 | |
0.2 mg/ml | |
Western Blot, ELISA, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Human | |
Q15124 | |
5239 | |
IgG1 κ | |
Protein A or G purified |
rPGM5/7164 | |
Western Blot 1-2 ug/ml, ELISA, Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin 2-4 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
10 mM PBS with 0.05% BSA | |
aciculin, EC 5.4.2.2, PGM-RP, PGMRPphosphoglucomutase-like protein 5, phosphoglucomutase 5, Phosphoglucomutase-related protein | |
Recombinant full-length human Phosphoglucomutase 5 protein (Uniprot: Q15124) | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title