Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospholamban Antibody, Biotin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297463B
Description
Phospholamban Polyclonal specifically detects Phospholamban in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Phospholamban | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
CMD1PPLBcardiac phospholamban, phospholamban | |
Produced in rabbits immunized with E. coli-derived Human Phospholamban fragment. | |
0.1 mL | |
Phospho Specific, Signal Transduction | |
5350 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Biotin | |
PBS | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction