Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospholipase A2 XII Antibody (7C7C9) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$548.00
Specifications
Antigen | Phospholipase A2 XII |
---|---|
Clone | 7C7C9 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, ELISA, CyTOF |
Description
Phospholipase A2 XII Monoclonal specifically detects Phospholipase A2 XII in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.Specifications
Phospholipase A2 XII | |
1 mg/mL | |
Western Blot, Flow Cytometry, ELISA, CyTOF | |
Unconjugated | |
RUO | |
Human | |
EC 3.1.1.4, group XII secreted phospholipase A2, group XIIA secreted phospholipase A2, group XIIA secretory phospholipase A2, GXII, GXII sPLA2, Phosphatidylcholine 2-acylhydrolase 12A, phospholipase A2, group XII, phospholipase A2, group XIIA, PLA2G12, ROSSY, sPLA2-XII | |
PLA2G12A | |
IgG1 | |
Affinity Purified |
7C7C9 | |
Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, CyTOF-ready | |
Monoclonal | |
Mouse | |
Lipid and Metabolism | |
PBS with 0.05% Sodium Azide | |
81579 | |
Purified recombinant fragment of human PLA2G12A (AA: 21-189) expressed in E. Coli. | |
Primary | |
21 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title