Learn More
Phospholipase B1 Antibody (OTI1B3), DyLight 755, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP273385IR
Catalog No. NBP273385IR
Only null left
Add to Cart
Description
Description
Phospholipase B1 Monoclonal specifically detects Phospholipase B1 in Human samples. It is validated for Western Blot.
Specifications
Specifications
Phospholipase B1
Monoclonal
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
PLB1
Full length recombinant protein of human Phospholipase B1 (NP_694566) produced in HEK293T cells.
0.1 mL
Primary
Human
IgG1
Western Blot
OTI1B3
Western Blot
FLJ30866, hPLB, phospholipase B1, PLBmembrane-associated
Mouse
Affinity Purified
Lipid and Metabolism
151056.0
Store at 4°C in the dark.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.