Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospholipase B1 Antibody (OTI1D3), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273386PE
Description
Phospholipase B1 Monoclonal antibody specifically detects Phospholipase B1 in Human samples. It is validated for Western BlotSpecifications
Phospholipase B1 | |
Monoclonal | |
PE | |
PBS | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot | |
OTI1D3 | |
Western Blot | |
FLJ30866, hPLB, phospholipase B1, PLBmembrane-associated | |
Full length human recombinant protein of human PLB1(NP_694566) produced in HEK293T cell. | |
0.1 mL | |
Lipid and Metabolism | |
151056 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction