Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospholipase C beta 2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$343.50 - $573.00
Specifications
Antigen | Phospholipase C beta 2 |
---|---|
Dilution | Western Blot 0.04 to 0.4 μg/mL, Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
Applications | Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Phospholipase C beta 2 Polyclonal antibody specifically detects Phospholipase C beta 2 in Human samples. It is validated for Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
Phospholipase C beta 2 | |
Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Lipid and Metabolism, Wnt Signaling Pathway | |
PBS, pH 7.2, 40% glycerol | |
5330 | |
IgG | |
Affinity purified |
Western Blot 0.04 to 0.4 μg/mL, Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
Polyclonal | |
Purified | |
RUO | |
Human | |
EC 3.1.4.11, FLJ38135, Phosphoinositide phospholipase C-beta-2,1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase beta-2, phospholipase C, beta 2, Phospholipase C-beta-2, PLC-beta-2 | |
This antibody was developed against a recombinant protein corresponding to the amino acids: EQISKMMELAREKQAAELKALKETSENDTKEMKKKLETKRLERIQGMTKVTTDKMAQERLKREINNSHIQEVVQVIKQMTEN | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title