Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospholipase C like 1 Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306275PCP
Description
Phospholipase C like 1 Polyclonal antibody specifically detects Phospholipase C like 1 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
Phospholipase C like 1 | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
but catalytically inactive protein, inactive phospholipase C-like protein 1, MGC126580, MGC138190, phospholipase C, epsilon, phospholipase C-deleted in lung carcinoma, phospholipase C-like 1, phospholipase C-related but catalytically inactive protein, PLCE, PLCL, PLC-L1, PRIP | |
Produced in rabbits immunized with E. coli-derived Human Phospholipase C like 1 fragment. | |
0.1 mL | |
Signal Transduction | |
5334 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
PerCP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction