Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospholipid Scramblase 1/PLSCR1 Antibody (SR1591), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32212250UL
Description
Phospholipid Scramblase 1/PLSCR1 Monoclonal antibody specifically detects Phospholipid Scramblase 1/PLSCR1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence, ImmunoprecipitationSpecifications
Phospholipid Scramblase 1/PLSCR1 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation | |
SR1591 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
Erythrocyte phospholipid scramblase, MmTRA1b, MMTRA1BCa(2+)-dependent phospholipid scramblase 1, phospholipid scramblase 1, PL scramblase 1 | |
A synthesized peptide derived from human phospholipid Scramblase 1/PLSCR1 (Uniprot #: O15162) | |
50 μg | |
Cardiovascular Biology, Neuroscience, Signal Transduction | |
5359 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction