Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphonoacetic acid, 98+%

Catalog No. aaa1211709
Click to view available options
Quantity:
10 g
25 g
100 g

CAS: 4408-78-0 | C2H5O5P | 140.03 g/mol

Phosphonoacetic acid inhibits deoxyribo nucleic acid polymerase, especially in viruses. It is used as an antiviral agent. Further, it is used in the preparation of 1-(dimethylamino)-2,2-bis[(dimethylimmonio)-methyl]ethene diperchlorate by reacting with phosphorus oxychloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phosphonoacetic acid inhibits deoxyribo nucleic acid polymerase, especially in viruses. It is used as an antiviral agent. Further, it is used in the preparation of 1-(dimethylamino)-2,2-bis[(dimethylimmonio)-methyl]ethene diperchlorate by reacting with phosphorus oxychloride.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4408-78-0
Molecular Formula C2H5O5P
Molecular Weight (g/mol) 140.03
MDL Number MFCD00004311
InChI Key XUYJLQHKOGNDPB-UHFFFAOYSA-N
Synonym phosphonoacetic acid, fosfonet, phosphonoacetate, acetic acid, phosphono, fosfonet sodium, phosphonacetic acid, carboxymethanephosphonic acid, fosfonoacetic acid, disodium phosphonoacetate, lopac-p-6909
PubChem CID 546
ChEBI CHEBI:15732
IUPAC Name 2-phosphonoacetic acid
SMILES OC(=O)CP(O)(O)=O

Specifications

Melting Point 139°C to 146°C
Odor Odorless
Quantity 10 g
UN Number UN3261
Beilstein 1764355
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 140.04
Percent Purity ≥98%
Chemical Name or Material Phosphonoacetic acid
Hazard Category Skin corrosion/irritation (category 1)
Hazard Statement Causes severe skin burns and eye damage
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 224-558-1
RTECSNumber AJ3278000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.