Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphoric Acid, 25% (v/v), Ricca Chemical
SDP

Catalog No. R58510001F Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R58510001F 1 L Cubitainer
585116 500 mL Natural Poly Bottle
585132 1 L Natural Poly Bottle
R585100020F 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. R58510001F Supplier Ricca Chemical Company Supplier No. 585132CT
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7732-18-5 , 7664-38-2
Molecular Formula H3O4P
Molecular Weight (g/mol) 97.994
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
Synonym orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico
PubChem CID 1004
ChEBI CHEBI:26078
IUPAC Name phosphoric acid
SMILES OP(=O)(O)O

Specifications

pH <2
Color Colorless
Melting Point ∼0°C
Boiling Point ∼100°C
CAS Min % 69.74, 30.27
CAS Max % 69.74, 30.27
Density 1.1866
Packaging Cubitainer
Quantity 1 L
Physical Form Liquid
Solubility Information Miscible in water
Percent Purity 25% V/V
Grade Laboratory
Chemical Name or Material Phosphoric Acid, 25% (v/v)
Show More Show Less
ShelfLife 24 months
Recommended Storage Ambient Corrosive
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.