Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphoric Acid, 40% v/v, AquaPhoenix Scientific™
SCI_ED

Catalog No. S88682 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S88682 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S88682 Supplier Aqua Phoenix Scientific Supplier No. PH7500P
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 7664-38-2 , 7732-18-5
Molecular Formula H3O4P
Molecular Weight (g/mol) 97.99
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
IUPAC Name phosphoric acid
SMILES OP(O)(O)=O

Specifications

Concentration 40% v/v
Quantity 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.