Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phosphoric Acid, 75% (w/w), Reagents
Click to view available options
Quantity:
2.5 L
4 L
55 gal.
Specifications
Specifications
pH | <2 |
Color | Colorless |
Boiling Point | 260°C |
Packaging | Poly Drum |
Specific Gravity | 1.579 |
Quantity | 55 gal. |
Physical Form | Liquid |
Percent Purity | 75% |
Grade | Reagent |
Chemical Name or Material | Phosphoric Acid, 75% (w/w) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction