Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphoric Acid ACS AR, Macron Fine Chemicals™

Catalog No. MK2796212X6 Shop All Avantor Products
Change view
Click to view available options
Quantity:
500 g
500 mL
2.5 kg
2.5 L
Packaging:
Glass
POLYSTORMOR
Poly
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MK-2796212X6 2.5 L Glass
MK-2816-500 500 g POLYSTORMOR
MK2816-212 2.5 kg Poly
MK-2796-500P 500 mL Poly
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. MK-2796212X6 Supplier Avantor Supplier No. 279646

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Chemical Identifiers

CAS 7664-38-2
Molecular Formula H3O4P
Molecular Weight (g/mol) 97.994
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
Synonym orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico
PubChem CID 1004
ChEBI CHEBI:26078
IUPAC Name phosphoric acid
SMILES OP(=O)(O)O

Specifications

Packaging Glass
Assay Percent Range 85%
Quantity 2.5 L
Formula Weight 98
Grade ACS
Chemical Name or Material Phosphoric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.