Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphoric acid, Puriss., meets analytical spec. of BP, FCC, NF, Ph. Eur., 85.0-88.0%, Solstice

Catalog No. 6001140 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
5 L
4 x 2.5 L
6 x 1 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-011-40 1 L
60-011-41 2.5 L
60-011-42 5 L
60-044-535 6 x 1 L
60-044-533 4 x 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 60-011-40 Supplier Solstice Supplier No. 041071L
Add to Cart
Add to Cart

Puriss., meets analytical specification of BP, FCC, NF, Ph. Eur., 85.0-88.0%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7664-38-2
Molecular Formula H3O4P
Molecular Weight (g/mol) 97.994
MDL Number MFCD00011340
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
Synonym orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico
PubChem CID 1004
ChEBI CHEBI:26078
IUPAC Name phosphoric acid
SMILES OP(=O)(O)O

Specifications

Concentration 85 to 88.0%
Density 1.685g/mL at 25°C
Vapor Pressure 5mmHg (25°C), 2.2mmHg (20°C)
Quantity 1 L
Linear Formula H3PO4
UN Number 1805
Beilstein 1921286
Formula Weight 98g/mol
Grade puriss.
Chemical Name or Material Phosphoric Acid, Meets analytical specification of Ph. Eur.BP, NF, FCC
Hazard Statement P280-P305 + P351 + P338-P310
Precautionary Statement H290-H314
EINECSNumber 231-633-2
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.