Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phosphoric Acid/Sulfuric Acid Solution, Reagents
Click to view available options
Quantity:
4 L
10 L
20 L
Specifications
Specifications
Chemical Name or Material | Phosphoric Acid/Sulfuric Acid Solution |
Color | Colorless |
CAS | 7732-18-5, 7664-93-9, 7664-38-2 |
Packaging | HDPE Winpak |
Specific Gravity | 1.2294 |
Quantity | 20 L |
Physical Form | Liquid |
Percent Purity | 15% |
Grade | Reagent |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction