Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphorous (III) Bromide, 99% metal basis, Alfa Aesar™

Click to view available options
Quantity:
25g
100g
7789-60-8
Br3P
270.686
MFCD00011436
IPNPIHIZVLFAFP-UHFFFAOYSA-N
phosphorus tribromide,phosphorous tribromide,tribromophosphine,phosphorus iii bromide,phosphorous bromide,pbr3,phosphorus bromide,phosphorus bromide pbr3,unii-58r3866pua,tribro-mophosphine
24614
tribromophosphane
P(Br)(Br)Br
7789-60-8
Br3P
270.686
MFCD00011436
IPNPIHIZVLFAFP-UHFFFAOYSA-N
phosphorus tribromide,phosphorous tribromide,tribromophosphine,phosphorus iii bromide,phosphorous bromide,pbr3,phosphorus bromide,phosphorus bromide pbr3,unii-58r3866pua,tribro-mophosphine
24614
tribromophosphane
P(Br)(Br)Br
Assay Percent Range 99% (Metals Basis)
UN Number 1808
Beilstein 3903060
Appearance Colorless liquid
Solubility Information Micible in acetone,carbon disulfide,hydrogen sulfide,chloroform,diethyl ether and decomposes in water and alcohol.
Formula Weight 270.7
Physical Form Liquid
Density 2.852 g/cm3 at 20°C
Odor Pungent
Refractive Index 1.697
Sensitivity Moisture sensitive
Boiling Point 172.9°C
Color Colorless
Melting Point -40°C
Quantity 100g
Chemical Name or Material Phosphorus(III) Bromide
Show More Show Less

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : DOT Class: 8, Packaging Group: II

EINECSNumber : 232-178-2

RTECSNumber : TH4460000

TSCA : Yes

Recommended Storage : Ambient temperatures