Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphorus(III) bromide, 99%

Catalog No. aa4019114
Click to view available options
Quantity:
25 g
100 g

CAS: 7789-60-8 | Br3P | 270.686 g/mol

Phosphorus(III) bromide is used mainly for the conversion of alcohols to alkyl bromides and it is used in tests for skin corrosivity. It is used for alkene preparation and it act as a lewis base and a lewis acid. It is used as a catalyst for the alpha-bromination of carboxylic acids and used as intermediates in hell-volhard-zelinsky halogenation. Phosphorus tribromide is used in the manufacture of pharmaceuticals such as alprazolam, methohexital and fenoprofen. It is also used as a fire suppression agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-60-8
Molecular Formula Br3P
Molecular Weight (g/mol) 270.686
MDL Number MFCD00011436
InChI Key IPNPIHIZVLFAFP-UHFFFAOYSA-N
Synonym phosphorus tribromide, phosphorous tribromide, tribromophosphine, phosphorus iii bromide, phosphorous bromide, pbr3, phosphorus bromide, phosphorus bromide pbr3, unii-58r3866pua, tribro-mophosphine
PubChem CID 24614
IUPAC Name tribromophosphane
SMILES P(Br)(Br)Br

Specifications

Boiling Point 172.9°C
Melting Point °C to 40°C
Color Colorless
Physical Form Liquid
Quantity 25 g
Assay Percent Range 99% (metals basis)
Beilstein 3903060
Solubility Information Micible in acetone,carbon disulfide,hydrogen sulfide,chloroform,diethyl ether and decomposes in water and alcohol.
Formula Weight 270.7
Odor Pungent
Refractive Index 1.697
Sensitivity Moisture sensitive
Appearance Colorless liquid
Density 2.852 g/cm3 at 20°C
Chemical Name or Material Phosphorus(III) bromide
Show More Show Less
Hazard Category H314-H335
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation DOT Class: 8, Packaging Group: II
EINECSNumber 232-178-2
RTECSNumber TH4460000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.