Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphorus AA Standard, 1000 ppm P in H2O, Ricca Chemical
SDP

Catalog No. p-3340430
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Natural LDPE Bottle
Natural Poly Bottle

Chemical Identifiers

CAS 7722-76-1, 7732-18-5
Molecular Formula H6NO4P
Molecular Weight (g/mol) 115.03
MDL Number MFCD00003396
InChI Key LFVGISIMTYGQHF-UHFFFAOYSA-N
PubChem CID 24402
ChEBI CHEBI:62982
IUPAC Name phosphoric acid amine
SMILES N.OP(O)(O)=O

Specifications

Packaging Natural LDPE Bottle
CAS Min % 98.0, 0.36
CAS Max % 100.0, 0.38
Physical Form Liquid
Color Colorless
Boiling Point 100°C
Type Standard
Quantity 100 mL
Concentration or Composition (by Analyte or Components) 1mL = 1mg P (1, 000ppm P)NH4H2PO4 in H2O
Chemical Name or Material Phosphorus AA Standard
ShelfLife 24 months
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.