Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphorus(III) chloride, 99.997% (metals basis), Thermo Scientific Chemicals

Catalog Number AA2298814
Click to view available options
Quantity:
25 g
100 g
7719-12-2
Cl3P
137.32
MFCD00011438
FAIAAWCVCHQXDN-UHFFFAOYSA-N
phosphorus trichloride, phosphorous trichloride, trichlorophosphine, phosphorus chloride, phosphorous chloride, phosphorus iii chloride, pcl3, phosphorus chloride pcl3, phosphine, trichloro, fosfortrichloride
24387
CHEBI:30334
trichlorophosphane
P(Cl)(Cl)Cl
7719-12-2
Cl3P
137.32
MFCD00011438
FAIAAWCVCHQXDN-UHFFFAOYSA-N
phosphorus trichloride, phosphorous trichloride, trichlorophosphine, phosphorus chloride, phosphorous chloride, phosphorus iii chloride, pcl3, phosphorus chloride pcl3, phosphine, trichloro, fosfortrichloride
24387
CHEBI:30334
trichlorophosphane
P(Cl)(Cl)Cl

Phosphorus(III) chloride is the precursor to organ phosphorus compounds, most notably phosphites and phosphonates. It is used during electrode position of metal on rubber and for making surfactants, gasoline additives, dyestuffs, textile finishing agents, medicinal products, and other chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 76°C
Melting Point -111.8°C
Physical Form Liquid
Quantity 25 g
UN Number UN1809
Merck Index 14,7358
Solubility Information Soluble in benzene,carbon sulfide,ether,chloroform,carbon tetrachloride.
Formula Weight 137.33
Percent Purity 99.997%
Odor Acrid
Refractive Index 1.516
Sensitivity Moisture sensitive
Density 1.574g/mL at 21°C
Assay (metals basis)
Chemical Name or Material Phosphorus(III) chloride
Show More Show Less
Hazard Category H300+H330-H314-H373
Hazard Statement GHS H Statement
H300-H330-H314-H318-H373
Fatal if swallowed.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: PHOSPHORUS TRICHLORIDE
EINECSNumber 231-749-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit