Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphorus(III) iodide, 99+%, Thermo Scientific™

Catalog No. ac221120500
Click to view available options
:
50g

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Specifications

CAS 13455-01-1
Quantity 50g
Assay Percent Range 99+%
Molecular Formula I3P
Linear Formula PI3
MDL Number MFCD00011442
Fieser 10,318
Synonym phosphorus triiodide, phosphorous triiodide, phosphorus iii iodide, triiodophosphine, phosphortrijodid, acmc-1bzov
InChI Key PZHNNJXWQYFUTD-UHFFFAOYSA-N
SMILES P(I)(I)I
IUPAC Name triiodophosphane
Molecular Weight (g/mol) 411.687
PubChem CID 83485
Formula Weight 411.69
Percent Purity ≥99.0%
Chemical Name or Material Phosphorus(III) iodide, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 236-647-2
TSCA TSCA
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.