Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphorus oxychloride, 99%, Thermo Scientific Chemicals

Catalog No. p-222019
Click to view available options
Quantity:
1 kg
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent, Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-87-3
MDL Number MFCD00011443
InChI Key XHXFXVLFKHQFAL-UHFFFAOYSA-N
Synonym phosphorus oxychloride, phosphoryl chloride, phosphoric trichloride, phosphoryl trichloride, phosphoroxychloride, trichlorophosphine oxide, phosphorus oxytrichloride, fosforoxychlorid, phosphorous oxychloride, phosphorylchlorid
PubChem CID 24813
ChEBI CHEBI:30336
SMILES O=P(Cl)(Cl)Cl

Specifications

Boiling Point 107.0°C
Melting Point 1.2°C
Physical Form Liquid
Quantity 1 kg
Assay Percent Range 98.5% min. (ex Cl) (Argentometry)
Molecular Formula Cl3OP
Linear Formula POCl3
Fieser 01,876; 02,330; 03,228; 04,390; 05,535; 07,292; 09,374; 11,429; 13,249; 15,267; 17,288
Merck Index 15, 7461
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in ccl4, chloroform and benzene
Molecular Weight (g/mol) 153.33
Formula Weight 153.33
Percent Purity 99%
Packaging Glass bottle
Refractive Index 1.4600 to 1.4620
Specific Gravity 1.645
Density 1.6450g/mL
Chemical Name or Material Phosphorus oxychloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Fatal if inhaled.
Causes damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Contact with water liberates toxic gas.
Reacts violently
Precautionary Statement GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF INHALED: Remove to fresh air a
EINECSNumber 233-046-7
RTECSNumber TH4897000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.