Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphorus pentoxide, ≥99.99% trace metals basis, MilliporeSigma™ Supelco™

Catalog No. 111018231
Click to view available options
Quantity:
50 g
250 g

Phosphorus pentoxide (P2O5) is a commonly used drying agent. It is a component of Eatons reagent, which is commonly used as a catalyst for dehydration reactions.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Synonym Phosphoric anhydride; Phosphorus(V) oxide

Specifications

Melting Point 340°C
CAS 1314-56-3
Quantity 250 g
Linear Formula P2O5
Formula Weight 141.94
Percent Purity ≥99.99% (trace metals basis)
Packaging Poly Bottle
Vapor Pressure 10 mmHg (238°C)
Density 2.3 g/mL (at 25°C) Vapor Density: 4.9 (vs air)
RTECSNumber TH3945000
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.