Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphorus pentoxide, 99+%, for analysis

Catalog No. AC215750025
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle

CAS: 1314-56-3 | O5P2 | 141.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Adsorbent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-56-3
Molecular Formula O5P2
Molecular Weight (g/mol) 141.94
MDL Number MFCD00011440
InChI Key DLYUQMMRRRQYAE-UHFFFAOYSA-N
Synonym Phosphoric anhydride
IUPAC Name tricyclo[3.3.1.1³,⁷]tetraphosphoxane-1,3,5,7-tetrone
SMILES O=P12OP3(=O)OP(=O)(O1)OP(=O)(O2)O3

Specifications

Melting Point 340.0°C to 360.0°C
Color White
Physical Form Very Deliquescing Powder
Quantity 2.5 kg
Assay Percent Range 99+%
Linear Formula P2O5
Fieser 01,871; 07,291; 10,319; 14,151; 15,267
Merck Index 15, 7467
Solubility Information Solubility in water: 85% (20°C); reacts forming h3po4 with evol.of heat
Formula Weight 141.94
Percent Purity ≥99%
Grade Analytical
Packaging Glass bottle
Density 2.39
Chemical Name or Material Phosphorus pentoxide, p.a.
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Reacts violently with water.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 215-236-1
RTECSNumber TH3945000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.