Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphorus(V) chloride, 99.998% (Metals basis), Alfa Aesar™

Click to view available options
Quantity:
25g
100g
10026-13-8
Cl5P
208.224
MFCD00011439
UHZYTMXLRWXGPK-UHFFFAOYSA-N
phosphorus pentachloride,phosphorus v chloride,pentachlorophosphorane,phosphorane, pentachloro,pcl5,phosphorous pentachloride,phosphorpentachlorid,phosphoric perchloride,phosphorus perchloride,unii-0ex753tydu
24819
CHEBI:30335
pentachloro-$l^{5}-phosphane
P(Cl)(Cl)(Cl)(Cl)Cl
10026-13-8
Cl5P
208.224
MFCD00011439
UHZYTMXLRWXGPK-UHFFFAOYSA-N
phosphorus pentachloride,phosphorus v chloride,pentachlorophosphorane,phosphorane, pentachloro,pcl5,phosphorous pentachloride,phosphorpentachlorid,phosphoric perchloride,phosphorus perchloride,unii-0ex753tydu
24819
CHEBI:30335
pentachloro-$l^{5}-phosphane
P(Cl)(Cl)(Cl)(Cl)Cl
Assay Percent Range 99.998% (metals basis)
Appearance Yellowish crystals
Solubility Information Insoluble in water.
Formula Weight 208.24
Physical Form Solid
Density 2.114 g/cm3 at 20°C
Odor Pungent
Sensitivity Moisture sensitive
Vapor Pressure 0.02 hPa at 20°C
Color Yellow
Melting Point 162° (sublimation)
Quantity 25g
Chemical Name or Material Phosphorus(V) chloride
Show More Show Less

Keep container tightly closed and dry. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).

P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302-H314-H330-H373

DOTInformation : DOT Class: 8, Packaging Group: II

EINECSNumber : 233-060-3

RTECSNumber : TB6125000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen